2-Cyclopropylpyridin-4-amine 95+%

Supplier: APOLLO SCIENTIFIC
Danger

OR306608-1G OR306608-250MG
77408-930EA 414.66 USD
77408-930 77408-932
2-Cyclopropylpyridin-4-amine 95+%
2-Cyclopropylpyridin-4-amine
Formula: C₈H₁₀N₂
MW: 134.178 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12032771
CAS Number: 340006-71-5
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR