3,6-Dibromo-1H-pyrazolo[4,3-c]pyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR300406-1G OR300406-250MG
77408-660EA 340.62 USD
77408-660 77408-662
3,6-Dibromo-1H-pyrazolo[4,3-c]pyridine
3,6-Dibromo-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₃Br₂N₃
MW: 276.916 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22380429
CAS Number: 1357946-34-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR