1-(5-Bromo-3-chloropyridin-2-yl)ethanone
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR300404-250MG
OR300404-1G
77408-654EA
96.78
USD
77408-654
77408-652
1-(5-Bromo-3-chloropyridin-2-yl)ethanone
1-(5-Bromo-3-chloropyridin-2-yl)ethanone
Formula:
C₇H₅BrClNO MW: 234.478 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18257815 CAS Number: 1256808-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...