3-Hydroxy-5-methylisoxazole 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Hymexazol

OR2998-25G OR2998-5G
77408-508EA 59.1 USD
77408-508 77408-510
3-Hydroxy-5-methylisoxazole 97%
3-Hydroxy-5-methylisoxazole
Formula: C₄H₅NO₂
MW: 99.09 g/mol
Melting Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00144468
CAS Number: 10004-44-1
EINECS: 233-000-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR