p-Toluenesulfonyl cyanide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Methylbenzenesulphonyl cyanide, Tosyl cyanide

OR46010-1G OR46010-25G OR46010-5G
77409-868EA 28.52 USD
77409-868 77409-870 77409-872
p-Toluenesulfonyl cyanide 97%
p-Toluenesulfonyl cyanide
Formula: C₈H₇NO₂S
Boiling Pt: 105…106 °C
Melting Pt: 47…50 °C
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00009977
CAS Number: 19158-51-1
EINECS: 242-849-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR