3-Iodobenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR460051-25G OR460051-5G
77409-860EA 325 USD
77409-860 77409-862
3-Iodobenzoyl chloride
3-Iodobenzoyl chloride
Formula: C₇H₄ClIO
Boiling Pt: 159…160 °C/23 mmHg
Melting Pt: 23…25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093934
CAS Number: 1711-10-0
EINECS: 216-979-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR