(2,6-Dichlorophenyl)methanesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-(Chlorosulphonyl)-2,6-dichlorotoluene

OR43513-1G OR43513-250MG
77409-698EA 268.96 USD
77409-698 77409-700
(2,6-Dichlorophenyl)methanesulfonyl chloride
(2,6-Dichlorophenyl)methanesulfonyl chloride
Formula: C₇H₅Cl₃O₂S
MW: 259.537 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11204033
CAS Number: 85952-31-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR