3-(Methylsulfonyl)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-3-(methylsulphonyl)benzene, 3-(Bromomethyl)phenyl methyl sulphone

OR40022-1G OR40022-250MG OR40022-5G
77409-448EA 228.22 USD
77409-448 77409-450 77409-452
3-(Methylsulfonyl)benzyl bromide 97%
3-(Methylsulfonyl)benzyl bromide
Formula: C₈H₉BrO₂S
MW: 249.125 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12025160
CAS Number: 82657-76-9
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR