2-(Bromomethyl)phenyl acetate

Supplier: APOLLO SCIENTIFIC
Danger

OR400175-1G OR400175-500MG OR400175-5G
77409-440EA 303.6 USD
77409-440 77409-442 77409-444
2-(Bromomethyl)phenyl acetate
2-(Bromomethyl)phenyl acetate
Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Storage Temperature: Freezer
MDL Number: MFCD20487652
CAS Number: 704-65-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR