1-Chloroisoquinoline-7-boronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Borono-1-chloroisoquinoline, 7-Borono-1-chloro-2-azanaphthalene, (1-Chloroisoquinolin-7-yl)boronic acid

OR300003-250MG OR300003-500MG
77408-514EA 505.73 USD
77408-514 77408-516 77408-518
1-Chloroisoquinoline-7-boronic acid
1-Chloroisoquinoline-7-boronic acid
Formula: C₉H₇BClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13190241
CAS Number: 370864-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR