3-Methylbenzofuran

Supplier: APOLLO SCIENTIFIC
Danger

OR27821-1G OR27821-5G
77407-994EA 59 USD
77407-994 77407-996
3-Methylbenzofuran
3-Methylbenzofuran
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 195…197 °C
Density: 1.046 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00205641
CAS Number: 21535-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR