1-Amino-8-bromoisoquinoline

Supplier: APOLLO SCIENTIFIC
Warning

OR300063-100MG OR300063-250MG OR300063-500MG
77408-538EA 173.54 USD
77408-538 77408-540 77408-542
1-Amino-8-bromoisoquinoline
1-Amino-8-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21365093
CAS Number: 1337879-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR