2-Chloroisonicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

OR29861-100G OR29861-1G OR29861-5G
77408-482EA 117.16 USD
77408-482 77408-484 77408-486
2-Chloroisonicotinic acid
2-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191402
CAS Number: 6313-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR