4-(4-Chlorophenoxy)aniline
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR29829-10G
OR29829-50G
77408-460EA
44.26
USD
77408-460
77408-462
4-(4-Chlorophenoxy)aniline
4-(4-Chlorophenoxy)aniline
Formula:
C₁₂H₁₀ClNO Boiling Pt: 205 °C/12 mmHg Melting Pt: 99…103 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043925 CAS Number: 101-79-1 EINECS: 202-976-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...