5-Methoxy-1,3-benzothiazol-2-amine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2965-5G OR2965-1G OR2965-25G
77408-420EA 258.78 USD
77408-420 77408-410 77408-412
5-Methoxy-1,3-benzothiazol-2-amine 98%
5-Methoxy-1,3-benzothiazol-2-amine
Formula: C₈H₈N₂OS
Storage Temperature: Refrigerator
MDL Number: MFCD00464314
CAS Number: 54346-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR