3,5-Dichlorobenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

OR29565-100G OR29565-25G OR29565-5G
77408-396EA 57.46 USD
77408-396 77408-398 77408-400
3,5-Dichlorobenzyl alcohol
3,5-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
Melting Pt: 76…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004634
CAS Number: 60211-57-6
EINECS: 262-109-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR