L-Phenylalanine ethyl ester hydrochloride 97%

Supplier: APOLLO SCIENTIFIC

OR28736-100G OR28736-25G
77408-200EA 44.82 USD
77408-200 77408-202
L-Phenylalanine ethyl ester hydrochloride 97%
L-Phenylalanine ethyl ester hydrochloride
Formula: C₁₁H₁₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00012507
CAS Number: 3182-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR