3-Bromo-6-chloro-2-methylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR2823-100G OR2823-25G OR2823-5G
77408-046EA 550.16 USD
77408-046 77408-048 77408-050
3-Bromo-6-chloro-2-methylpyridine
3-Bromo-6-chloro-2-methylpyridine
Formula: C₆H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD07375117
CAS Number: 132606-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR