4-Mercaptobenzoic acid 90%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Thiobenzoic acid

OR28166-10G OR28166-25G OR28166-5G
77408-036EA 114.1 USD
77408-036 77408-038 77408-040
4-Mercaptobenzoic acid 90%
4-Mercaptobenzoic acid
Formula: C₇H₆O₂S
MW: 154.1863 g/mol
Melting Pt: 215…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016617
CAS Number: 1074-36-8
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR