Metacetamol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-(3-Hydroxyphenyl)acetamide, N-Acetyl-3-aminophenol, Paracetamol, 3-Acetamidophenol, 3'-Hydroxyacetanilide

OR27724-100G OR27724-25G OR27724-5G
77407-970EA 110.04 USD
77407-970 77407-972 77407-974
Metacetamol
Metacetamol
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002263
CAS Number: 621-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR