(Bromomethyl)cyclobutane 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR451132-1G OR451132-25G OR451132-5G
77409-766EA 21.36 USD
77409-766 77409-768 77409-770
(Bromomethyl)cyclobutane 98%
(Bromomethyl)cyclobutane
Formula: C₅H₉Br
Boiling Pt: 145…148 °C
Density: 1.339 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00040960
CAS Number: 17247-58-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR