4-(Bromomethyl)phenyl acetate 97%, tech

Supplier: APOLLO SCIENTIFIC
Danger

OR400140-1G OR400140-250MG OR400140-5G
77409-428EA 206.82 USD
77409-428 77409-430 77409-432
4-(Bromomethyl)phenyl acetate 97%, tech
4-(Bromomethyl)phenyl acetate
Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17013576
CAS Number: 52727-95-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR