1-(Tributylstannyl)-2,3-dimethylbenzene

Supplier: APOLLO SCIENTIFIC

Synonyms: tributyl-(2,3-dimethylphenyl)stannane

OR399040-250MG OR399040-1G
77409-414EA 103.92 USD
77409-414 77409-412
1-(Tributylstannyl)-2,3-dimethylbenzene
1-(Tributylstannyl)-2,3-dimethylbenzene
Formula: C₂₀H₃₆Sn
Storage Temperature: Ambient
CAS Number: 1229597-06-1
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR