2-Amino-5-chloropyridine-3-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR346130-1G OR346130-250MG OR346130-5G
77409-266EA 215.98 USD
77409-266 77409-268 77409-270
2-Amino-5-chloropyridine-3-sulfonyl chloride
2-Amino-5-chloropyridine-3-sulfonyl chloride
Formula: C₅H₄Cl₂N₂O₂S
MW: 227.07 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD12402514
CAS Number: 163137-45-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR