6-Bromo-2-chloro-3-iodopyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR345567-1G OR345567-25G OR345567-5G
77409-258EA 66.22 USD
77409-258 77409-260 77409-262
6-Bromo-2-chloro-3-iodopyridine 97%
6-Bromo-2-chloro-3-iodopyridine
Formula: C₅H₂BrClIN
Storage Temperature: Ambient
MDL Number: MFCD11857690
CAS Number: 1138444-17-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR