1-Butyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Butyl isothiocyanate

OR30240-100G OR30240-25G OR30240-500G
77408-838EA 45.84 USD
77408-838 77408-840 77408-842
1-Butyl isothiocyanate
1-Butyl isothiocyanate
Formula: C₅H₉NS
Boiling Pt: 63…65 °C/11 mmHg
Density: 0.954 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00004824
CAS Number: 592-82-5
EINECS: 209-770-4
UN: 2927
ADR: 6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR