3-Bromo-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR301082-10G OR301082-1G OR301082-5G
77408-762EA 83.54 USD
77408-762 77408-764 77408-766
3-Bromo-1H-pyrazole
3-Bromo-1H-pyrazole
Formula: C3H3BrN2
MW: 146.97 g/mol
Boiling Pt: 70…72 °C/0.1 mmHg
Melting Pt: 60…65 °C
Storage Temperature: Freezer
MDL Number: MFCD01646049
CAS Number: 14521-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR