tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR301080-1G OR301080-5G OR301080-10G
77408-754EA 124.3 USD
77408-754 77408-756 77408-752
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate 97%
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate
Formula: C11H20N2O2
Storage Temperature: Ambient
MDL Number: MFCD17016700
CAS Number: 201162-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR