6-(Dimethoxymethyl)-2-thionicotinonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-(Dimethoxymethyl)-2-sulphanylpyridine-3-carbonitrile

OR29716-10G OR29716-1G OR29716-5G
77408-424EA 1682.88 USD
77408-424 77408-426 77408-428
6-(Dimethoxymethyl)-2-thionicotinonitrile
6-(Dimethoxymethyl)-2-thionicotinonitrile
Formula: C₉H₁₀N₂O₂S
MW: 210.253 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173693
CAS Number: 175277-23-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR