Tri(2-thienyl)phosphine

Supplier: APOLLO SCIENTIFIC
Warning

OR29178-1G OR29178-25G OR29178-5G
77408-352EA 29.54 USD
77408-352 77408-354 77408-356
Tri(2-thienyl)phosphine
Tri(2-thienyl)phosphine
Formula: C₁₂H₉PS₃
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00015471
CAS Number: 24171-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR