1-Acetyl-6-amino-2,3-dihydro-(1H)-indole 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR2669-1G OR2669-5G
77407-828EA 65.2 USD
77407-828 77407-830
1-Acetyl-6-amino-2,3-dihydro-(1H)-indole 95+%
1-Acetyl-6-amino-2,3-dihydro-(1H)-indole
Formula: C₁₀H₁₂N₂O
Melting Pt: 182…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056021
CAS Number: 62368-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR