4-(4-Methoxyphenyl)-1-butanol 95%

Supplier: APOLLO SCIENTIFIC

OR2659-100G OR2659-10G OR2659-25G
77407-818EA 325 USD
77407-818 77407-820 77407-822
4-(4-Methoxyphenyl)-1-butanol 95%
4-(4-Methoxyphenyl)-1-butanol
Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00002972
CAS Number: 52244-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR