2-(3-Bromo-5-chlorophenyl)acetonitrile 95%

Supplier: APOLLO SCIENTIFIC

OR31036-250MG OR31036-5G OR31036-1G
77409-014EA 41.78 USD
77409-014 77409-016 77409-012
2-(3-Bromo-5-chlorophenyl)acetonitrile 95%
2-(3-Bromo-5-chlorophenyl)acetonitrile
Formula: C₈H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD13195464
CAS Number: 1056454-88-6
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR