3,5-Dimethyl-1-phenyl-1H-pyrazole 97+%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3,5-Dimethyl-1-phenylpyrazole, 3,5-Dimethyl-1-phenyl-1H-pyrazole

OR300627-1G OR300627-5G
77408-676EA 28.52 USD
77408-676 77408-678
3,5-Dimethyl-1-phenyl-1H-pyrazole 97+%
3,5-Dimethyl-1-phenyl-1H-pyrazole
Formula: C₁₁H₁₂N₂
MW: 172.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020728
CAS Number: 1131-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR