2-Methoxypyridine-3,4-diamine

Supplier: APOLLO SCIENTIFIC
Warning

OR300373-1G OR300373-500MG
77408-602EA 481.9 USD
77408-602 77408-604
2-Methoxypyridine-3,4-diamine
2-Methoxypyridine-3,4-diamine
Formula: C₆H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD10574697
CAS Number: 33631-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR