7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one

Supplier: APOLLO SCIENTIFIC
Warning

OR300314-1G OR300314-250MG
77408-594EA 1372.46 USD
77408-594 77408-596
7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one
7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one
Formula: C₁₄H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD29765459
CAS Number: 1301219-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR