2-(4-Chlorophenoxy)isobutyric acid 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR28848-25G OR28848-500G
77408-264EA 108 USD
77408-264 77408-266
2-(4-Chlorophenoxy)isobutyric acid 97+%
2-(4-Chlorophenoxy)isobutyric acid
Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00004192
CAS Number: 882-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR