4-[(4-Chlorophenyl)sulfanyl]thiophene-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Carboxythien-3-yl 4-chlorophenyl sulphide, 4-[(4-Chlorophenyl)sulphanyl]-3-thenoic acid

OR25411-1G OR25411-250MG OR25411-5G
77407-596EA 190.52 USD
77407-596 77407-598 77407-600
4-[(4-Chlorophenyl)sulfanyl]thiophene-3-carboxylic acid
4-[(4-Chlorophenyl)sulfanyl]thiophene-3-carboxylic acid
Formula: C₁₁H₇ClO₂S₂
Melting Pt: 195…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052149
CAS Number: 149609-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR