6-Nitroquinoline

Supplier: APOLLO SCIENTIFIC

OR2346-25G OR2346-100G
77407-334EA 48.9 USD
77407-334 77407-332
6-Nitroquinoline
6-Nitroquinoline
Formula: C₉H₆N₂O₂
Melting Pt: 151…153 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00006799
CAS Number: 613-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR