Isopropyl 2-bromobenzoate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Prop-2-yl 2-bromobenzoate
OR26849-1G
OR26849-5G
77407-862EA
21.26
USD
77407-862
77407-864
Isopropyl 2-bromobenzoate
Isopropyl 2-bromobenzoate
Formula:
C₁₀H₁₁BrO₂ MW: 243.097 g/mol Boiling Pt: 92 °C (4 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00027951 CAS Number: 59247-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...