1,8-Octanediol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,8-Dihydroxy octane

OR2431-100G OR2431-1KG OR2431-500G
77407-466EA 63.16 USD
77407-466 77407-468 77407-470
1,8-Octanediol 98%
1,8-Octanediol
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 279
Melting Pt: 59…61 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002989
CAS Number: 629-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR