2-Morpholino-3-pyridinamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Amino-2-morpholin-4-ylpyridine

OR23393-100MG OR23393-10G OR23393-1G
77407-312EA 20.4 USD
77407-312 77407-314 77407-316
2-Morpholino-3-pyridinamine
2-Morpholino-3-pyridinamine
Formula: C₉H₁₃N₃O
Melting Pt: 127…128 °C
Storage Temperature: Ambient
MDL Number: MFCD03001243
CAS Number: 51627-47-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR