4-(1H-Pyrazol-1-yl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR23288-1G OR23288-250MG OR23288-5G
77407-216EA 23.36 USD
77407-216 77407-218 77407-220
4-(1H-Pyrazol-1-yl)benzoic acid
4-(1H-Pyrazol-1-yl)benzoic acid
Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
Melting Pt: 263…272 °C
Storage Temperature: Ambient
MDL Number: MFCD02682056
CAS Number: 16209-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR