2-Bromothiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR23133-100G OR23133-25G OR23133-500G
77407-124EA 23.4 USD
77407-124 77407-126 77407-128
2-Bromothiazole
2-Bromothiazole
Formula: C₃H₂BrNS
MW: 164.03 g/mol
Boiling Pt: 171 °C
Density: 1.82 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005316
CAS Number: 3034-53-5
EINECS: 221-229-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR