Methanesulfonic acid 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR322096-100G OR322096-500G
77409-148EA 24.46 USD
77409-148 77409-150
Methanesulfonic acid 99%
Methanesulphonic acid
Formula: CH₃SO₃H
MW: 96.11 g/mol
Boiling Pt: 167 °C/10 mmHg
Melting Pt: 17…19 °C
Density: 1.481 g/cm³ (25 °C)
Flash Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR