2-Picolyl bromide hydrobromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Bromomethyl)pyridine hydrobromide

OR322016-5G OR322016-1G OR322016-25G
77409-134EA 36.68 USD
77409-134 77409-130 77409-132
2-Picolyl bromide hydrobromide 95%
2-Picolyl bromide hydrobromide
Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD01863544
CAS Number: 31106-82-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR