1,8-Dibromonaphthalene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR315403-100G OR315403-25G OR315403-5G
77409-064EA 376.96 USD
77409-064 77409-066 77409-068
1,8-Dibromonaphthalene 98%
1,8-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00183574
CAS Number: 17135-74-9
UN: 3077
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR