(S)-(+)-2-Methylbutyric acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR315294-1G OR315294-250MG OR315294-5G
77409-054EA 117.16 USD
77409-054 77409-056 77409-058
(S)-(+)-2-Methylbutyric acid 98%
(S)-(+)-2-Methylbutyric acid
Formula: C₅H₁₀O₂
Boiling Pt: 78 °C/15 mmHg
Density: 0.938 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063166
CAS Number: 1730-91-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR