8-Bromopyrido[4,3-b]pyrazine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 8-Bromopyrido[3,4-b]pyrazine

OR30605-1G OR30605-250MG
77408-906EA 353.52 USD
77408-906 77408-908
8-Bromopyrido[4,3-b]pyrazine
8-Bromopyrido[4,3-b]pyrazine
Formula: C₇H₄BrN₃
MW: 210.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834590
CAS Number: 929074-45-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR