6-Bromo-o-toluic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR3054-1G OR3054-25G OR3054-5G
77408-896EA 21.34 USD
77408-896 77408-898 77408-900
6-Bromo-o-toluic acid 97%
6-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 107…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01310788
CAS Number: 90259-31-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR